Product Details
- SNP ID
-
rs200112350
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75206129 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGGGCCAGGAGCCGGGGACGGC[A/G]TCCTCCCCATTCACGATCCTGGACA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTRB2
PubMed Links
Gene Details
- Gene
- CTRB2
- Gene Name
- chymotrypsinogen B2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001025200.3 |
150 |
Silent Mutation |
GAC,GAT |
D39D |
NP_001020371.3 |
View Full Product Details