Product Details

SNP ID
rs200966797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57512817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCGGCCTCCTCGGTGCCAAAG[C/T]GAGCACTGCGGATCTTCCCGAAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC102A PubMed Links

Gene Details

Gene
CCDC102A
Gene Name
coiled-coil domain containing 102A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033212.3 1790 Missense Mutation CAC,CGC H526R NP_149989.2
XM_011523469.2 1790 Missense Mutation CAC,CGC H526R XP_011521771.1

View Full Product Details