Product Details

SNP ID
rs200478905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67664002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGGGTGGTGGGAGGCCAGGGC[C/T]GCAGCGGGAGTGCGCCCGCAGGAAG
Phenotype
MIM: 609377 MIM: 610859 MIM: 607484
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACD PubMed Links

Gene Details

Gene
ACD
Gene Name
adrenocortical dysplasia homolog
There are no transcripts associated with this gene.

Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
CARMIL2
Gene Name
capping protein regulator and myosin 1 linker 2
There are no transcripts associated with this gene.

Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
PARD6A
Gene Name
par-6 family cell polarity regulator alpha
There are no transcripts associated with this gene.

View Full Product Details