Product Details

SNP ID
rs201161071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81041437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGTCCAATTGAAGGCTGCCCC[A/T]GAGGCCCTGAGAGACCGTTTTCTCA
Phenotype
MIM: 614693 MIM: 611509
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATMIN PubMed Links

Gene Details

Gene
ATMIN
Gene Name
ATM interactor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300728.1 376 UTR 5 NP_001287657.1
NM_015251.2 376 Nonsense Mutation AGA,TGA R140* NP_056066.2
Gene
CENPN
Gene Name
centromere protein N
There are no transcripts associated with this gene.

View Full Product Details