Product Details

SNP ID
rs200349448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20418157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCTGGGGAAGCAGTCCAGGAA[A/G]GCAGCCAATGTGCTGGGGGGTGCAT
Phenotype
MIM: 614361
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSM5 PubMed Links

Gene Details

Gene
ACSM5
Gene Name
acyl-CoA synthetase medium-chain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324371.1 Intron NP_001311300.1
NM_001324372.1 Intron NP_001311301.1
NM_001324373.1 Intron NP_001311302.1
NM_017888.2 Intron NP_060358.2
XM_006721056.3 Intron XP_006721119.1

View Full Product Details