Product Details

SNP ID
rs201130844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72060526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGTTATGTTTCTGGCTGGGGGC[A/G]AAATGCCAATTTTAAATTTACTGAC
Phenotype
MIM: 140100 MIM: 140210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HP PubMed Links

Gene Details

Gene
HP
Gene Name
haptoglobin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126102.2 741 Missense Mutation CAA,CGA Q227R NP_001119574.1
NM_001318138.1 741 Missense Mutation CAA,CGA Q227R NP_001305067.1
NM_005143.4 741 Missense Mutation CAA,CGA Q286R NP_005134.1
Gene
HPR
Gene Name
haptoglobin-related protein
There are no transcripts associated with this gene.

Gene
TXNL4B
Gene Name
thioredoxin like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142317.1 741 Intron NP_001135789.1
NM_001142318.1 741 Intron NP_001135790.1
NM_001324354.1 741 Intron NP_001311283.1
NM_001324355.1 741 Intron NP_001311284.1
NM_017853.2 741 Intron NP_060323.1
XM_017023377.1 741 Intron XP_016878866.1

View Full Product Details