Product Details

SNP ID
rs199893447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3284170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGAAGCGGTGACCCTTGTGGAG[C/G]ATATGCAGAGAGAGCTTGGGAGACT
Phenotype
MIM: 604191
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF263 PubMed Links

Gene Details

Gene
ZNF263
Gene Name
zinc finger protein 263
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005741.4 411 Missense Mutation CAT,GAT H118D NP_005732.2
XM_006720831.1 411 Missense Mutation CAT,GAT H118D XP_006720894.1
XM_006720832.1 411 Missense Mutation CAT,GAT H118D XP_006720895.1
XM_006720833.1 411 Missense Mutation CAT,GAT H118D XP_006720896.1
XM_006720834.3 411 UTR 5 XP_006720897.1
XM_011522344.1 411 Missense Mutation CAT,GAT H118D XP_011520646.1
XM_017022850.1 411 UTR 5 XP_016878339.1

View Full Product Details