Product Details

SNP ID
rs199859922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1945516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTCACCATGAAGGCCCTCTTCT[C/G]TTGGGCTGTCTGGAAGCGGCCATGG
Phenotype
MIM: 606216
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MSRB1 PubMed Links

Gene Details

Gene
MSRB1
Gene Name
methionine sulfoxide reductase B1
There are no transcripts associated with this gene.

Gene
RPL3L
Gene Name
ribosomal protein L3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005061.2 1657 Missense Mutation CAG,GAG Q384E NP_005052.1
XM_011522571.2 1657 Missense Mutation CAG,GAG Q389E XP_011520873.1

View Full Product Details