Product Details

SNP ID
rs201234837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56962052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGCTCCAAAGGCACCTCGCAC[A/G]AGGCAGGCATCGTGTGCCGCATCAC
Phenotype
MIM: 118470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CETP PubMed Links

Gene Details

Gene
CETP
Gene Name
cholesteryl ester transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000078.2 90 Missense Mutation AAG,GAG K25E NP_000069.2
NM_001286085.1 90 Missense Mutation AAG,GAG K25E NP_001273014.1
XM_006721124.3 90 Missense Mutation AAG,GAG K25E XP_006721187.1

View Full Product Details