Product Details

SNP ID
rs200195038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30006869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGATGCCTACCAGCAGTCCCCGG[C/T]GCCGCGAGCTGTAGCTGAGACTCAG
Phenotype
MIM: 604567
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOC2A PubMed Links

Gene Details

Gene
DOC2A
Gene Name
double C2 domain alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282062.1 986 Missense Mutation CAC,CGC H265R NP_001268991.1
NM_001282063.1 986 Missense Mutation CAC,CGC H265R NP_001268992.1
NM_001282068.1 986 Missense Mutation CAC,CGC H265R NP_001268997.1
NM_003586.2 986 Missense Mutation CAC,CGC H265R NP_003577.2
XM_011545975.1 986 Missense Mutation CAC,CGC H265R XP_011544277.1
XM_017023776.1 986 Missense Mutation CAC,CGC H265R XP_016879265.1
XM_017023777.1 986 Missense Mutation CAC,CGC H265R XP_016879266.1
XM_017023778.1 986 Missense Mutation CAC,CGC H147R XP_016879267.1
Gene
INO80E
Gene Name
INO80 complex subunit E
There are no transcripts associated with this gene.

View Full Product Details