Product Details
- SNP ID
-
rs200426119
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:48167581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGACGAGCACGGTGCAGCCCTGG[A/G]AGGCTTCACGGATTGTGCGCTGGAT
- Phenotype
-
MIM: 607040
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ABCC11
PubMed Links
Gene Details
- Gene
- ABCC11
- Gene Name
- ATP binding cassette subfamily C member 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032583.3 |
4321 |
Missense Mutation |
TCC,TTC |
S1324F |
NP_115972.2 |
NM_033151.3 |
4321 |
Missense Mutation |
TCC,TTC |
S1324F |
NP_149163.2 |
NM_145186.2 |
4321 |
Missense Mutation |
TCC,TTC |
S1286F |
NP_660187.1 |
XM_011523397.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1005F |
XP_011521699.1 |
XM_011523398.2 |
4321 |
Missense Mutation |
TCC,TTC |
S701F |
XP_011521700.1 |
XM_017023795.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1324F |
XP_016879284.1 |
XM_017023796.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1324F |
XP_016879285.1 |
XM_017023797.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1324F |
XP_016879286.1 |
XM_017023798.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1324F |
XP_016879287.1 |
XM_017023799.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1324F |
XP_016879288.1 |
XM_017023800.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1324F |
XP_016879289.1 |
XM_017023801.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1288F |
XP_016879290.1 |
XM_017023802.1 |
4321 |
Missense Mutation |
TCC,TTC |
S1005F |
XP_016879291.1 |
XM_017023803.1 |
4321 |
Intron |
|
|
XP_016879292.1 |
View Full Product Details