Product Details

SNP ID
rs202230194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69341288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAATACTTCTCAGTACCAGGGC[A/G]TGGTGGTGTACTCCATGGCAGACAT
Phenotype
MIM: 606979
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG8 PubMed Links

Gene Details

Gene
COG8
Gene Name
component of oligomeric golgi complex 8
There are no transcripts associated with this gene.

Gene
NIP7
Gene Name
NIP7, nucleolar pre-rRNA processing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199434.1 709 Intron NP_001186363.1
NM_016101.4 709 Missense Mutation ATG,GTG M131V NP_057185.1
Gene
TMED6
Gene Name
transmembrane p24 trafficking protein 6
There are no transcripts associated with this gene.

View Full Product Details