Product Details

SNP ID
rs202000368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23679226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGCCGCACCAGAGGGAGAAGAT[A/G]TCCATGGAAATATCCATTCACCGCA
Phenotype
MIM: 612962 MIM: 602098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCTN5 PubMed Links

Gene Details

Gene
DCTN5
Gene Name
dynactin subunit 5
There are no transcripts associated with this gene.

Gene
PLK1
Gene Name
polo like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005030.5 Intron NP_005021.2

View Full Product Details