Product Details

SNP ID
rs202041463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3284188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGAGGATATGCAGAGAGAGCTT[C/G]GGAGACTGAGACAACAGGTGAGAGA
Phenotype
MIM: 604191
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF263 PubMed Links

Gene Details

Gene
ZNF263
Gene Name
zinc finger protein 263
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005741.4 429 Missense Mutation CGG,GGG R124G NP_005732.2
XM_006720831.1 429 Missense Mutation CGG,GGG R124G XP_006720894.1
XM_006720832.1 429 Missense Mutation CGG,GGG R124G XP_006720895.1
XM_006720833.1 429 Missense Mutation CGG,GGG R124G XP_006720896.1
XM_006720834.3 429 UTR 5 XP_006720897.1
XM_011522344.1 429 Missense Mutation CGG,GGG R124G XP_011520646.1
XM_017022850.1 429 UTR 5 XP_016878339.1

View Full Product Details