Product Details

SNP ID
rs202141965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4425904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGGCTGCGCGGTGCTCCACAC[A/G]CTGGTTGCTGGTGGTTGTGGGGACC
Phenotype
MIM: 611668 MIM: 608382
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CORO7 PubMed Links

Gene Details

Gene
CORO7
Gene Name
coronin 7
There are no transcripts associated with this gene.

Gene
CORO7-PAM16
Gene Name
CORO7-PAM16 readthrough
There are no transcripts associated with this gene.

Gene
DNAJA3
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135110.2 100 Missense Mutation CAC,CGC H8R NP_001128582.1
NM_001286516.1 100 UTR 5 NP_001273445.1
NM_005147.5 100 Missense Mutation CAC,CGC H8R NP_005138.3

View Full Product Details