Product Details

SNP ID
rs199766569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8075687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTCCACGGCTGCGTCACTCGGG[C/T]AATAATAGGTGATGATCTCCGTCAC
Phenotype
MIM: 603741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALOX12B PubMed Links

Gene Details

Gene
ALOX12B
Gene Name
arachidonate 12-lipoxygenase, 12R type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139.2 1823 Missense Mutation TAC,TGC Y521C NP_001130.1

View Full Product Details