Product Details

SNP ID
rs200144448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40390114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAAGCTGGATCCCTTTTAGTTC[C/T]TTTTGGGGCAGCCCTTTTTTTGGCA
Phenotype
MIM: 126430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOP2A PubMed Links

Gene Details

Gene
TOP2A
Gene Name
topoisomerase (DNA) II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001067.3 4419 Missense Mutation AGA,GGA R1440G NP_001058.2
XM_005257632.1 4419 Missense Mutation AGA,GGA R1428G XP_005257689.1
XM_011525165.1 4419 Intron XP_011523467.1

View Full Product Details