Product Details

SNP ID
rs200279652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40093142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGAATGCATGTTGTTCAGGGTCC[A/G]CAGGTCCGGCAGCTTGAGCAGCAGC
Phenotype
MIM: 602408 MIM: 190120
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR1D1 PubMed Links

Gene Details

Gene
NR1D1
Gene Name
nuclear receptor subfamily 1 group D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021724.4 1808 Missense Mutation CGG,TGG R596W NP_068370.1
Gene
THRA
Gene Name
thyroid hormone receptor, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190918.1 1808 Silent Mutation CCA,CCG P372P NP_001177847.1
NM_001190919.1 1808 Silent Mutation CCA,CCG P411P NP_001177848.1
NM_003250.5 1808 Silent Mutation CCA,CCG P411P NP_003241.2
NM_199334.3 1808 Intron NP_955366.1

View Full Product Details