Product Details

SNP ID
rs200391097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29043246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCATGGCGGCTCAGAAAGATCT[C/G]TGGGACGCCATTGTGATTGGGGCGG
Phenotype
MIM: 616713
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PIPOX PubMed Links

Gene Details

Gene
PIPOX
Gene Name
pipecolic acid and sarcosine oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016518.2 347 Silent Mutation CTC,CTG L7L NP_057602.2
XM_017024722.1 347 Intron XP_016880211.1

View Full Product Details