Product Details

SNP ID
rs199719387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75899583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCTTGAAGAGCAGGAAGGCAA[A/G]ACGGTGGATGCCGGAGCCTCGGGCA
Phenotype
MIM: 616807 MIM: 611844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBF1 PubMed Links

Gene Details

Gene
FBF1
Gene Name
Fas binding factor 1
There are no transcripts associated with this gene.

Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032478.3 1027 Missense Mutation CTT,TTT L268F NP_115867.2
Gene
TRIM65
Gene Name
tripartite motif containing 65
There are no transcripts associated with this gene.

View Full Product Details