Product Details
- SNP ID
-
rs199855472
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41487432 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGATGATCTCCGTCTGGCAGCACTG[C/T]AGCTGCTCCGAGCTGGACACCACCT
- Phenotype
-
MIM: 602764
MIM: 604540
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT35
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2301354] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT35
- Gene Name
- keratin 35
There are no transcripts associated with this gene.
- Gene
- KRT36
- Gene Name
- keratin 36
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003771.4 |
906 |
Silent Mutation |
CTA,CTG |
L302L |
NP_003762.1 |
View Full Product Details