Product Details

SNP ID
rs200511075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3198187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTATGTTCAGGTCTTTTCCAC[A/G]GTCTTCCAAGTTCCATCTACCAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1A2 PubMed Links

Gene Details

Gene
OR1A2
Gene Name
olfactory receptor family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012352.2 671 Silent Mutation ACA,ACG T223T NP_036484.1

View Full Product Details