Product Details

SNP ID
rs199652449
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8345415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTCGAGTTGGAAAGGAATGTAT[C/T]CATCCCCATAGGCTGGAGCTATTTC
Phenotype
MIM: 610097
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ODF4 PubMed Links

Gene Details

Gene
ODF4
Gene Name
outer dense fiber of sperm tails 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319953.1 709 Missense Mutation TCC,TTC S61F NP_001306882.1
NM_153007.4 709 Missense Mutation TCC,TTC S176F NP_694552.2
XM_005256476.3 709 Missense Mutation TCC,TTC S72F XP_005256533.1
XM_011523688.1 709 Missense Mutation TCC,TTC S72F XP_011521990.1

View Full Product Details