Product Details

SNP ID
rs199686657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28737477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTCTGCATCCGTGCCCTCCTC[A/G]ACCTCCACACCGACGTGGGCAGTGT
Phenotype
MIM: 609799 MIM: 602464
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEK8 PubMed Links

Gene Details

Gene
NEK8
Gene Name
NIMA related kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178170.2 994 Missense Mutation AAC,GAC N264D NP_835464.1
XM_011524638.2 994 Missense Mutation AAC,GAC N288D XP_011522940.1
XM_011524640.2 994 Missense Mutation AAC,GAC N288D XP_011522942.1
XM_017024499.1 994 Missense Mutation AAC,GAC N264D XP_016879988.1
XM_017024500.1 994 Missense Mutation AAC,GAC N225D XP_016879989.1
XM_017024501.1 994 Missense Mutation AAC,GAC N288D XP_016879990.1
Gene
TLCD1
Gene Name
TLC domain containing 1
There are no transcripts associated with this gene.

Gene
TRAF4
Gene Name
TNF receptor associated factor 4
There are no transcripts associated with this gene.

View Full Product Details