Product Details

SNP ID
rs200338233
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47733796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTACGCCGCCCCGGACCCGCG[C/T]GCCGGGCTCTACCCGGGGCCGCGTG
Phenotype
MIM: 604895
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TBX21 PubMed Links

Gene Details

Gene
TBX21
Gene Name
T-box 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013351.1 557 Silent Mutation CGC,CGT R114R NP_037483.1
XM_011524698.2 557 Silent Mutation CGC,CGT R114R XP_011523000.1

View Full Product Details