Product Details
- SNP ID
-
rs200550508
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:15741970 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTACAGCCCAAATACTGCCTGGCT[C/T]GAGAGGCTCCTATCGCACCAGGAGC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TBC1D26
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17855672] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TBC1D26
- Gene Name
- TBC1 domain family member 26
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178571.4 |
945 |
Missense Mutation |
TCG,TTG |
S232L |
NP_848666.2 |
View Full Product Details