Product Details

SNP ID
rs201539562
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47953088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTCCAGGCGGGTGTTAAAGTC[C/G]CTGTTGGGGCCTCCAGCGTCAGGCC
Phenotype
MIM: 603287
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PNPO PubMed Links

Gene Details

Gene
PNPO
Gene Name
pyridoxamine 5'-phosphate oxidase
There are no transcripts associated with this gene.

Gene
PRR15L
Gene Name
proline rich 15 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024320.3 427 Silent Mutation AGC,AGG S49R NP_077296.1
XM_005257663.4 427 Silent Mutation AGC,AGG S49R XP_005257720.1
XM_005257664.2 427 Silent Mutation AGC,AGG S49R XP_005257721.1
XM_005257665.1 427 Silent Mutation AGC,AGG S49R XP_005257722.1
XM_005257666.4 427 Silent Mutation AGC,AGG S49R XP_005257723.1

View Full Product Details