Product Details

SNP ID
rs201846516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75131097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGGGGGCTGGTGCAGATGAAG[A/G]CCTGCGTGCTGCGGAGAAGGACGCG
Phenotype
MIM: 191720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMC7 PubMed Links

Gene Details

Gene
ARMC7
Gene Name
armadillo repeat containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304271.1 372 Intron NP_001291200.1
NM_001304272.1 372 Intron NP_001291201.1
NM_001304273.1 372 Intron NP_001291202.1
NM_024585.3 372 Intron NP_078861.1
Gene
HN1
Gene Name
hematological and neurological expressed 1
There are no transcripts associated with this gene.

Gene
NT5C
Gene Name
5', 3'-nucleotidase, cytosolic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252377.1 372 Missense Mutation GCC,GTC A95V NP_001239306.1
NM_014595.2 372 Missense Mutation GCC,GTC A95V NP_055410.1
XM_011524700.2 372 Intron XP_011523002.2

View Full Product Details