Product Details

SNP ID
rs201510371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42189771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGAAGGACCCGGGGCATCGCAG[A/G]GGGGAAGGGGGAGGAGGGGCAGGCA
Phenotype
MIM: 608587 MIM: 602358 MIM: 604528 MIM: 604260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GHDC PubMed Links

Gene Details

Gene
GHDC
Gene Name
GH3 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142623.1 1851 UTR 3 NP_001136095.1
NM_032484.4 1851 Missense Mutation CCT,TCT P509S NP_115873.1
Gene
HCRT
Gene Name
hypocretin neuropeptide precursor
There are no transcripts associated with this gene.

Gene
KCNH4
Gene Name
potassium voltage-gated channel subfamily H member 4
There are no transcripts associated with this gene.

Gene
STAT5B
Gene Name
signal transducer and activator of transcription 5B
There are no transcripts associated with this gene.

View Full Product Details