Product Details

SNP ID
rs201443807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27761164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACTTCCGCACAAAGCAGGGCAC[C/T]GGGTCTTGGGGCTTCAGGCTGTTGA
Phenotype
MIM: 163730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOS2 PubMed Links

Gene Details

Gene
NOS2
Gene Name
nitric oxide synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000625.4 2729 Silent Mutation CCA,CCG P956P NP_000616.3
XM_011524859.2 2729 Silent Mutation CCA,CCG P956P XP_011523161.1
XM_011524860.2 2729 Silent Mutation CCA,CCG P955P XP_011523162.1
XM_011524861.1 2729 Silent Mutation CCA,CCG P932P XP_011523163.1
XM_011524862.1 2729 Silent Mutation CCA,CCG P734P XP_011523164.1

View Full Product Details