Product Details

SNP ID
rs201649537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3566877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCGTCCTCTGGCTTCAGAGACC[C/T]TGAAAACTGTCGCAGATAAACTTCC
Phenotype
MIM: 602076 MIM: 607066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPV1 PubMed Links

Gene Details

Gene
TRPV1
Gene Name
transient receptor potential cation channel subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018727.5 2659 Missense Mutation AGG,GGG R820G NP_061197.4
NM_080704.3 2659 Missense Mutation AGG,GGG R820G NP_542435.2
NM_080705.3 2659 Missense Mutation AGG,GGG R820G NP_542436.2
NM_080706.3 2659 Missense Mutation AGG,GGG R820G NP_542437.2
Gene
TRPV3
Gene Name
transient receptor potential cation channel subfamily V member 3
There are no transcripts associated with this gene.

View Full Product Details