Product Details

SNP ID
rs200794627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28328721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTACTTTACACAAAGCTTCATA[C/T]TCAACCCGATACCTGAAAAACAAAA
Phenotype
MIM: 614394 MIM: 612912 MIM: 191161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT20 PubMed Links

Gene Details

Gene
IFT20
Gene Name
intraflagellar transport 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267774.1 482 Silent Mutation GAA,GAG E136E NP_001254703.1
NM_001267775.1 482 Silent Mutation GAA,GAG E110E NP_001254704.1
NM_001267776.1 482 Silent Mutation GAA,GAG E110E NP_001254705.1
NM_001267777.1 482 Intron NP_001254706.1
NM_001267778.1 482 UTR 3 NP_001254707.1
NM_174887.3 482 UTR 3 NP_777547.1
Gene
TMEM97
Gene Name
transmembrane protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014573.2 482 Intron NP_055388.2
XM_005257965.3 482 Intron XP_005258022.1
Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
There are no transcripts associated with this gene.

View Full Product Details