Product Details

SNP ID
rs201551910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50375381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGATTGTGAAGCCTCCTGTCCTC[C/T]AGCACCAGAGTTATTTTCACCACCT
Phenotype
MIM: 610885 MIM: 614854 MIM: 611837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EME1 PubMed Links

Gene Details

Gene
EME1
Gene Name
essential meiotic structure-specific endonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166131.1 251 Missense Mutation CCA,CTA P58L NP_001159603.1
NM_152463.2 251 Missense Mutation CCA,CTA P58L NP_689676.2
XM_005257081.3 251 Missense Mutation CCA,CTA P58L XP_005257138.1
XM_011524395.2 251 UTR 5 XP_011522697.1
XM_017024236.1 251 Missense Mutation CCA,CTA P58L XP_016879725.1
Gene
LRRC59
Gene Name
leucine rich repeat containing 59
There are no transcripts associated with this gene.

Gene
MRPL27
Gene Name
mitochondrial ribosomal protein L27
There are no transcripts associated with this gene.

View Full Product Details