Product Details

SNP ID
rs201212813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42782761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCCGTCCCCCATCCCACAGACT[C/T]GGAAACTGTCTAGAGCTGAGCGGCA
Phenotype
MIM: 610907 MIM: 601844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPS25 PubMed Links

Gene Details

Gene
VPS25
Gene Name
vacuolar protein sorting 25 homolog
There are no transcripts associated with this gene.

Gene
WNK4
Gene Name
WNK lysine deficient protein kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321299.1 690 UTR 5 NP_001308228.1
NM_032387.4 690 Missense Mutation CGG,TGG R208W NP_115763.2
XM_011525135.2 690 Missense Mutation CGG,TGG R208W XP_011523437.1
XM_017024962.1 690 Missense Mutation CGG,TGG R208W XP_016880451.1
XM_017024963.1 690 Missense Mutation CGG,TGG R208W XP_016880452.1
XM_017024964.1 690 Missense Mutation CGG,TGG R208W XP_016880453.1
XM_017024965.1 690 Missense Mutation CGG,TGG R208W XP_016880454.1
XM_017024966.1 690 Intron XP_016880455.1

View Full Product Details