Product Details

SNP ID
rs201898433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7236370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCAGCTCCTGGCCCCAACTTT[C/T]GGTTCTTTCGGTCCTTCTTTCGAGG
Phenotype
MIM: 610684 MIM: 602151 MIM: 605125 MIM: 612910
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTDNEP1 PubMed Links

Gene Details

Gene
CTDNEP1
Gene Name
CTD nuclear envelope phosphatase 1
There are no transcripts associated with this gene.

Gene
DVL2
Gene Name
dishevelled segment polarity protein 2
There are no transcripts associated with this gene.

Gene
GABARAP
Gene Name
GABA type A receptor-associated protein
There are no transcripts associated with this gene.

Gene
PHF23
Gene Name
PHD finger protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284517.1 583 Missense Mutation NP_001271446.1
NM_001284518.1 583 Missense Mutation NP_001271447.1
NM_024297.2 583 Missense Mutation NP_077273.2

View Full Product Details