Product Details

SNP ID
rs202244473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7236469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGCCGGGAGGTATGTGTCAGG[A/G]ATGTGGGGGACAAAGGAGATGCCAC
Phenotype
MIM: 610684 MIM: 602151 MIM: 605125 MIM: 612910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTDNEP1 PubMed Links

Gene Details

Gene
CTDNEP1
Gene Name
CTD nuclear envelope phosphatase 1
There are no transcripts associated with this gene.

Gene
DVL2
Gene Name
dishevelled segment polarity protein 2
There are no transcripts associated with this gene.

Gene
GABARAP
Gene Name
GABA type A receptor-associated protein
There are no transcripts associated with this gene.

Gene
PHF23
Gene Name
PHD finger protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284517.1 484 Missense Mutation TCC,TTC S86F NP_001271446.1
NM_001284518.1 484 Missense Mutation TCC,TTC S149F NP_001271447.1
NM_024297.2 484 Missense Mutation TCC,TTC S153F NP_077273.2

View Full Product Details