Product Details

SNP ID
rs201083633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43028553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGGCCTCCCTTGGCCGTGGCC[A/G]TAGGCAGCTGCGCCGAACTGACTCA
Phenotype
MIM: 601555 MIM: 604631
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RND2 PubMed Links

Gene Details

Gene
RND2
Gene Name
Rho family GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005440.4 504 Missense Mutation CAT,CGT H186R NP_005431.1
XM_011525316.1 504 Missense Mutation CAT,CGT H186R XP_011523618.1
XM_011525317.2 504 Missense Mutation CAT,CGT H158R XP_011523619.1
Gene
VAT1
Gene Name
vesicle amine transport 1
There are no transcripts associated with this gene.

View Full Product Details