Product Details

SNP ID
rs201460712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1635205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTGGTCACAGGGCCCGAGCTCTC[A/C]TGGACGCTGCCCTCAATGGCTTCCA
Phenotype
MIM: 607873 MIM: 610791
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCARF1 PubMed Links

Gene Details

Gene
SCARF1
Gene Name
scavenger receptor class F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003693.3 1039 Missense Mutation CAG,CAT Q682H NP_003684.2
NM_145350.2 1039 UTR 3 NP_663325.1
XM_017025256.1 1039 Missense Mutation CAG,CAT Q271H XP_016880745.1
Gene
SLC43A2
Gene Name
solute carrier family 43 member 2
There are no transcripts associated with this gene.

View Full Product Details