Product Details

SNP ID
rs201494482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7353347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCACCGACTCTGAGTGTCTAGG[G/T]GCTTTGCGGGCCCGATTTGGTGTGG
Phenotype
MIM: 607763 MIM: 609848
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACAP1 PubMed Links

Gene Details

Gene
ACAP1
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014716.3 1459 Intron NP_055531.1
Gene
KCTD11
Gene Name
potassium channel tetramerization domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002914.2 1459 Silent Mutation GGG,GGT G135G NP_001002914.1
Gene
TMEM95
Gene Name
transmembrane protein 95
There are no transcripts associated with this gene.

View Full Product Details