Product Details

SNP ID
rs201906189
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30563324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGACTCCTCCAAGGGTGCCA[G/T]GACAACAAGCCTTGAGCCGGGGAGA
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
TBC1D29 PubMed Links

Gene Details

Gene
TBC1D29
Gene Name
TBC1 domain family member 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015594.2 501 Nonsense Mutation GGA,TGA G118* NP_056409.1
XM_011524596.1 501 Nonsense Mutation GGA,TGA G118* XP_011522898.1

View Full Product Details