Product Details

SNP ID
rs200778781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80997015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACCCTCGTCCACAGGTGATGTC[A/C]ATTGAAGAGGTGGAGAGGATCCTGG
Phenotype
MIM: 610901
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHMP6 PubMed Links

Gene Details

Gene
CHMP6
Gene Name
charged multivesicular body protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024591.4 435 Silent Mutation TCA,TCC S119S NP_078867.2
XM_005257668.1 435 Silent Mutation TCA,TCC S119S XP_005257725.1
Gene
LOC100129503
Gene Name
uncharacterized LOC100129503
There are no transcripts associated with this gene.

View Full Product Details