Product Details

SNP ID
rs201362915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56985381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTCTTCTCTCTTCCATAGGGC[A/G]GTCCAGGCGGTTCTAGCACTGGATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCPEP1 PubMed Links

Gene Details

Gene
SCPEP1
Gene Name
serine carboxypeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021626.2 264 Missense Mutation AGT,GGT S77G NP_067639.1
XM_005257557.2 264 Missense Mutation AGT,GGT S77G XP_005257614.1

View Full Product Details