Product Details
- SNP ID
-
rs202189623
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:34285874 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTGCCTTCAGCCCCCAGGGGCT[C/T]GCTGGGCCAGGTAAGCCCCCCAACT
- Phenotype
-
MIM: 601156
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCL11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1129844] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCL11
- Gene Name
- C-C motif chemokine ligand 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002986.2 |
207 |
Silent Mutation |
CTC,CTT |
L22L |
NP_002977.1 |
View Full Product Details