Product Details

SNP ID
rs200145735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49562364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCCTCTGCTATTGCTTTGCTG[C/T]GGGGAGCCCCGTACCTTTTGGTCCA
Phenotype
MIM: 603684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPG PubMed Links

Gene Details

Gene
LIPG
Gene Name
lipase G, endothelial type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308006.1 334 Missense Mutation GCG,GTG A19V NP_001294935.1
NM_006033.3 334 Missense Mutation GCG,GTG A19V NP_006024.1
XM_005258390.1 334 Intron XP_005258447.1
XM_011526265.2 334 Intron XP_011524567.1
XM_011526267.1 334 Intron XP_011524569.1
XM_017026095.1 334 Intron XP_016881584.1

View Full Product Details