Product Details

SNP ID
rs200482387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:7231638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGCCGACCTCTCTAGACTGAGGA[A/C]GCTAAACCTCAGCAACAACTTCTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRRC30 PubMed Links

Gene Details

Gene
LRRC30
Gene Name
leucine rich repeat containing 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105581.1 500 Missense Mutation AAG,ACG K167T NP_001099051.1

View Full Product Details