Product Details

SNP ID
rs200072155
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5891747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGACCGGTTTTTGCTGCCACTG[C/T]TACTGCTGTTGGCCGTGGTTGGGAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TMEM200C PubMed Links

Gene Details

Gene
TMEM200C
Gene Name
transmembrane protein 200C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080209.1 358 Missense Mutation AAC,AGC N106S NP_001073678.1
XM_005258130.4 358 Missense Mutation AAC,AGC N106S XP_005258187.1
XM_011525729.2 358 Missense Mutation AAC,AGC N106S XP_011524031.1

View Full Product Details