Product Details

SNP ID
rs201536919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:14752987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGGCAACACTGCTGTCCTATG[A/G]TGCAGTCATCGAGGTGCAAAACAAG
Phenotype
MIM: 616565
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD30B PubMed Links

Gene Details

Gene
ANKRD30B
Gene Name
ankyrin repeat domain 30B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145029.1 570 Missense Mutation GAT,GGT D162G NP_001138501.1
XM_006722325.3 570 Intron XP_006722388.1
XM_011525662.2 570 Missense Mutation GAT,GGT D162G XP_011523964.1
XM_011525664.2 570 Intron XP_011523966.1
XM_011525665.2 570 Missense Mutation GAT,GGT D162G XP_011523967.1
XM_011525666.2 570 Missense Mutation GAT,GGT D162G XP_011523968.1

View Full Product Details