Product Details

SNP ID
rs200335256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57398710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGATTGGATACCTTCATTTGG[C/G]AAGAACCACAGAGTTCACATGGCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF548 PubMed Links

Gene Details

Gene
ZNF548
Gene Name
zinc finger protein 548
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172773.1 709 Silent Mutation GGC,GGG G153G NP_001166244.1
NM_152909.3 709 Silent Mutation GGC,GGG G141G NP_690873.2
XM_017026316.1 709 Silent Mutation GGC,GGG G144G XP_016881805.1

View Full Product Details