Product Details
- SNP ID
-
rs199848394
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48661696 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACGTCTAGTGCTCCGGCCTGGGA[C/T]TGGGTGTGGGTGCCCGCACGCCGCG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NTN5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs281392] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NTN5
- Gene Name
- netrin 5
- Gene
- SEC1P
- Gene Name
- secretory blood group 1, pseudogene
There are no transcripts associated with this gene.
View Full Product Details