Product Details

SNP ID
rs199675550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54307431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGTTCACCTTCCAGCTGCAGC[C/T]TGGGGGCTTCTCTGGCTGTGCCAAT
Phenotype
MIM: 606047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LILRA5 PubMed Links

Gene Details

Gene
LILRA5
Gene Name
leukocyte immunoglobulin like receptor A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021250.3 Intron NP_067073.1
NM_181879.2 Intron NP_870994.1
NM_181985.3 Intron NP_871714.1
NM_181986.2 Intron NP_871715.1

View Full Product Details